Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MEPE Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8689R-CY5.5
BOSSBS-8689R-CY5.5EA 900 EUR InStock
BOSSBS-8689R-CY5.5
Anti-MEPE Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
MEPE is a 525 amino acid extracellular matrix protein. Expressed in osteocytes and brain, MEPE is a regulator of bone metabolism that is thought to mediate mineralization and demineralization within the osteocyte microenvironment. MEPE contains an RGD cell-attachment motif and shares molecular similarities with several dentin-bone extracellular matrix RGD-containing phosphoglycoproteins, including OPN (osteopontin) and DSP (dentin sialophosphoprotein). Via its ability to control bone mineralization, MEPE is associated with various developmental events such as skeletogenesis, bone regeneration and odontogenesis. MEPE is secreted in hypophosphatemic osteomalacia tumors, suggesting a possible role in the pathophysiology of bone-related cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MEPE
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge