Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GJC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8697R-FITC
BOSSBS-8697R-FITCEA 900 EUR InStock
BOSSBS-8697R-FITC
Anti-GJC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Connexin 29 belongs to the connexin family and is a member of the epsilon-type subfamily. Connexin 29 is a membrane bound, multi-pass protein also known as gap junction epsilon-1 protein. A connexon, consisting of connexin hexamers, is a membrane bound structure that is integral in the formation of a gap junction. One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low molecular weight diffuse from one cell to a neighboring cell. Connexin 29 expression is restricted to the central nervous system and is present in brain, spinal cord, and sciatic nerve samples. It has been suggested that connexin 29 in the mature CNS contributes minimally to gap junctional intercellular communication in oligodendrocyte cell bodies. Rather, connexin 29 is targeted to myelin where it, along with connexin 32, may contribute to connexin-mediated communication between adjacent layers of uncompacted myelin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Connexin-29
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge