Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Viperin Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8718R
BOSSBS-8718REA 725 EUR InStock
BOSSBS-8718R
Anti-Viperin Rabbit Polyclonal Antibody
Antikörper
RSAD2 is a 361 amino acid protein that is involved in antiviral defense against pathogens such as Hep C, Cytomegalovirus and HIV-1. Localized to the cytosolic side of the endoplasmic reticulum and relocated to the Golgi apparatus upon viral infection, RSAD2 is thought to prevent viral budding by disrupting lipid rafts at the plasma membrane and supporting the Interferon-induced antiviral state of the cell. Additionally, RSAD2 can bind to and inactivate FDPS (an enzyme that is crucial for the synthesis of cholesterol and geranylated and farnesylated proteins), thereby playing a role in lipid synthesis. Overexpression of RSAD2 leads to abnormal lipid accumulation that is associated with atherosclerosis, a chronic inflammatory disease characterized by hardened arteries.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Viperin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge