Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NOB1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8857R-CY5
BOSSBS-8857R-CY5EA 900 EUR InStock
BOSSBS-8857R-CY5
Anti-NOB1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
NOB1P is a 412 amino acid nuclear protein that is involved in proteosome biogenesis and is required for the final step in 18S rRNA maturation. NOB1P contains a PIN domain, which functions as a nuclease in nonsense-mediated mRNA decay and is required for pre-rRNA cleavage. NOB1P interacts with Rent2, which is involved in nonsense-mediated decay of mRNAs containing premature stop codons. Expressed in placenta, spleen, endothelial cells, liver and lung, NOB1P is essential for the synthesis of 40S ribosome subunits. Supression of the gene encoding NOB1P inhibits the processing of the 20S pre-rRNA to the mature 18S rRNA, therefore leading to accumulation of high levels of 20S pre-rRNA with degradation intermediates.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge