Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KPNA2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8864R-CY5
BOSSBS-8864R-CY5EA 900 EUR InStock
BOSSBS-8864R-CY5
Anti-KPNA2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Protein transport across the nucleus is a selective, multi-step process involving several cytoplasmic factors that mediate protein passage through the nuclear pore complex (NPC). Cytoplasmic proteins that contain nuclear localization signals (NLSs) must be recognized as import substrates, dock at the nuclear pore complex and translocate across the nuclear envelope in an ATP-dependent fashion. Karyopherin alpha 1 and karyopherin alpha 6 are widely expressed nuclear import proteins that act as adaptors for karyopherin ∫1, specifically binding to and guiding NLS-containing proteins to the NPC. Both karyopherin alpha 1 and karyopherin alpha 6 contain one IBB domain and ten ARM repeats through which they convey their protein binding and localization function. Together, karyopherin å1 and karyopherin å6 are responsible for ensuring the nuclear import of NLS-containing substrates

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KPNA2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge