Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PISD Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8866R-CY7
BOSSBS-8866R-CY7EA 900 EUR InStock
BOSSBS-8866R-CY7
Anti-PISD Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Enzymes known as phosphatidylserine decarboxylases (PSDs) catalyze the formation of phosphatidylethanolamine from phosphatidylserine via phosphatidylserine decarboxylation. Type I PSDs contain LGST motifs and are found in bacteria and eukaryotic mitochondria, whereas type II PSDs contain GGST motifs and are found in eukaryotic endomembrane systems. PISD (phosphatidylserine decarboxylase), also known as phosphatidylserine decarboxylase proenzyme, PSDC, PSD, PSSC, DJ858B16, dJ858B16.2 or DKFZp566G2246, is a 408 amino acid a type I phosphatidylserine decarboxylase that localizes to the inner mitochondrial membrane. PISD contains a conserved LGST motif which is cleaved to produce two isoforms known as PISD α and PISD β. PISD is capable of forming a heterodimer and is highly expressed in liver and testis. The gene encoding PISD maps to human chromosome 22q12.2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PISD
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge