Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ORC1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8998R-FITC
BOSSBS-8998R-FITCEA 900 EUR InStock
BOSSBS-8998R-FITC
Anti-ORC1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The initiation of DNA replication is a multi-step process that depends on the formation of pre-replication complexes, which trigger initiation (1). Among the proteins required for establishing these complexes are the origin recognition complex (ORC) proteins (1). ORC proteins bind specifically to origins of replication where they serve as scaffold for the assembly of additional initiation factors (1). Human ORC subunits 1-6 are expressed in the nucleus of proliferating cells and tissues, such as the testis (2). ORC1 and ORC2 are both expressed at equivalent concentrations throughout the cell cycle; however, only ORC2 remains stably bound to chromatin (3,4). ORC4 and ORC6 are also expressed constantly throughout the cell cycle (5,6). ORC2, ORC3, ORC4 and ORC5 form a core complex upon which ORC6 and ORC1 assemble (7,8). The formation of this core complex suggests that ORC proteins play a crucial role in the G1-S transition in mammalian cells (8).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ORC1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge