Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RCBTB1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9007R-CY7
BOSSBS-9007R-CY7EA 900 EUR InStock
BOSSBS-9007R-CY7
Anti-RCBTB1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The BTB domain, also known as the POZ (poxvirus and zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. RCBTB1 (regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1), also known as GLP, CLLD7, CLLL7 or E4.5, is a 531 amino acid protein that localizes to the nucleus and contains two BTB (POZ) domains and six RCC1 repeats. Expressed ubiquitously, RCBTB1 is thought to be involved in cell cycle regulation, specifically via chromatin remodeling. The gene encoding RCBTB1 maps to a region on human chromosome 13 that is frequently deleted in B-cell chronic lymphocytic leukemia, suggesting a possible role for RCBTB1 in tumor suppression.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RCBTB1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge