Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DPP6 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9010R-CY5.5
BOSSBS-9010R-CY5.5EA 900 EUR InStock
BOSSBS-9010R-CY5.5
Anti-DPP6 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
DPP6 is a Type-II serine proteinase of the clan SC. The clan SC proteinases have a catalytic triad of Ser-Asp-His, and like other Serine proteinases, the active site serine is in a Gly-Xaa-Ser-Xaa -Gly orientation. DPP6 has an Asp instead of Ser in the catalytic site. DPP6 is a member of a broader family of dipeptidyl peptidases including DPP4, FAP/Seprase, DPP2, DPP8, DPP9, DPP10, which have differing substrate specificity and tissue localizations. The surface-bound DPP6 is a homodimer, and cleavage of in the stalk region releases a shed form of DPP6. The shed is the form found in serum. DPP6 has been found in highest abundance in the brain, but also in the kidney, liver and lung.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DPP6
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge