Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EFHD1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9015R-CY5.5
BOSSBS-9015R-CY5.5EA 900 EUR InStock
BOSSBS-9015R-CY5.5
Anti-EFHD1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The EF-hand domain is a twelve amino acid loop motif that is commonly found in proteins that participate in calcium-binding events within the cell. EF-hand domains generally exist in a pair that, together, form a stable four-helix bundle that enables the binding of calcium ions. Swiprosin-2, also known as EFHD1 (EF-hand domain-containing protein D1), SWS2, PP3051 or MST133, is a 239 amino acid protein that contains two EF-hand domains and is expressed in a wide variety of tissues, including brain, liver, heart, kidney, testis, ovaries and spleen. Expression of Swiprosin-2, a possible calcium-binding protein, is upregulated during neuronal differentiation, suggesting a role for Swiprosin-2 in brain development and maturation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EFHD1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge