Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PDXK Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9035R-A488
BOSSBS-9035R-A488EA 893 EUR InStock
BOSSBS-9035R-A488
Anti-PDXK Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Pyridoxal kinase belongs to the pyridoxine kinase family and phosphorylates vitamin B6, a step necessary for the conversion of vitamin B6 to pyridoxal 5’ phosphate (PLP), the active form of vitamin B6. PLP acts as a coenzyme and functions to maintain homeostasis. Pyridoxal kinase is a 312-amino acid cytoplasmic protein that may act as a homodimer and is expressed ubiquitously. There are three known isoforms of pyridoxal kinase, and isoform 3 expression is observed in adult testis and spermatozoa. The optimum pH for pyridoxal kinase is between 5.5 and 6.0. PDXK, the gene that encodes the pyridoxal kinase protein, maps to chromosome 21q22.3 and may be a candidate gene for autoimmune polyglandular disease type 1, a genetic disorder that has been mapped to the same region on chromosome 21.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDXK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge