Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PDZD7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9042R-FITC
BOSSBS-9042R-FITCEA 900 EUR InStock
BOSSBS-9042R-FITC
Anti-PDZD7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
PDZK7, also known as PDZD7, is a 517 amino acid protein that contains two PDZ (DHR) domains. Encoded by a gene that maps to human chromosome 10q24.31, PDZK7 is conserved in dog, mouse and rat, and exists as three alternatively spliced isoforms. PDZK7 is known to interact with Harmonin, MASS1, USH1G and Usherin. Localizing to nucleus, PDZK7 is expressed in retinal pigment epithelium and inner ear. Biallelic inactivation of PDZK7 can cause non-syndromic hearing impairment and chromosomal aberrations, which are linked to non-syndromic sensorineural deafness. PDZK7 mutations are also linked to Usher syndrome, which is characterized by retinitis pigmentosa and sensorineural deafness, and Alzheimer disease. The gene that encodes PDZK7 maps to human chromosome 10q24.31.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDZD7
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge