Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DIS3L2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9053R-CY3
BOSSBS-9053R-CY3EA 900 EUR InStock
BOSSBS-9053R-CY3
Anti-DIS3L2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The exosome is a multisubunit complex composed of several highly conserved subunits, some of which are 3’ to 5’ exoribonucleases. The complex is involved in a variety of cellular processes and is responsible for degrading unstable mRNAs that contain AU-rich (ARE) elements in their untranslated 3’ region. DIS3L2 (DIS3-like exonuclease 2) is an 885 amino acid protein that is thought to function as an exonuclease and may be required for the 3’ processing of pre-mRNA into mature mRNA. Defects or chromosomal translocations involving the gene encoding DIS3L2 may be associated with Marfanoid habitus, a genetic disorder characterized by abnormalities in the skeleton, eyes and cardiovascular system. DIS3L2 is expressed as five isoforms due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIS3L2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge