Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DRAP1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9055R-CY7
BOSSBS-9055R-CY7EA 900 EUR InStock
BOSSBS-9055R-CY7
Anti-DRAP1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
DR1, also known as NC2∫ (negative cofactor 2 subunit ∫), is a TFIID (TATA box-binding protein)-associated protein. DR1 localizes to the nucleus and contains an N-terminal histone fold motif, a TFIID-binding domain and an alanine and glutamine rich region. Via its histone fold motif, DR1 forms a heterodimer with NC2å (DRAP1) to comprise the conserved eukaryotic complex, NC2 (negative cofactor 2). The NC2 complex can both positively and negatively regulate transcription by RNA Pol II. More specifically, NC2 acts as a repressor of TATA-dependent transcription and acts as an activator for DPE-dependent transcription. NC2 represses RNA Pol II transcription by binding to TFIID and inhibiting association of the transcription factors TFIIA and TFIIB. NC2 activity is regulated by phosphorylation. Both subunits, NC2å and DR1, are phosphorylated in vivo.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DRAP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge