Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ODZ3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9061R-A680
BOSSBS-9061R-A680EA 893 EUR InStock
BOSSBS-9061R-A680
Anti-ODZ3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Teneurin-3, also known as Ten-3, TNM3 or ODZ3, is a 2,699 amino acid single-pass type II membrane protein that contains 25 YD repeats, 8 EGF-like domains, 5 NHL repeats and one teneurin N-terminal domain. Localised to the membrane and expressed in brain, testis and ovary, Teneurin-3 exists as a disulfide-liked homodimer that is thought to function as a cellular signal transducer. Additionally, Teneurin-3 may participate in eye-specific patterning in the visual pathway and is required for aligned binocular vision. The gene encoding Teneurin-3 maps to chromosome 4. Representing approximately 6% of the human genome, chromosome 4 contains nearly 900 genes, one of which is the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Type: Primary
Antigen: ODZ3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1721-1850/2699
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge