Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HIP12 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9067R-A680
BOSSBS-9067R-A680EA 893 EUR InStock
BOSSBS-9067R-A680
Anti-HIP12 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Huntington disease is associated with the expansion of a polyglutamine tract, greater than 35 repeats, in the HD gene product, huntingtin. HIP1, a membrane-associated protein, binds specifically to the N-terminus of human huntingtin. HIP1 is ubiquitously expressed in different brain regions at low levels and exhibits nearly identical subcellular fractionation as huntingtin. The HIP1 gene locates to the human chromosome 7q11.23. The huntingtin-HIP1 interaction is restricted to the brain and is inversely correlated to the polyglutamine length in the huntingtin, suggesting that loss of normal huntingtin-HIP1 interaction may compromise the membrane-cytoskeletal integrity in the brain. HIP1 contains an endocytic multidomain protein with a C-terminal Actin-binding domain, a central coiled-coil forming region and an N-terminal ENTH domain. HIP1 may be involved in vesicle trafficking; the structural integrity of HIP1 is crucial for maintenance of normal vesicle sise in vivo. HIP12 is a non-proapoptotic member of the HIP gene family that is expressed in the brain and shares a similar subcellular distribution pattern with HIP1. However, HIP12 differs from HIP1 in its pattern of expression at both the mRNA and protein level. HIP12 does not directly interact with huntingtin but can interact with HIP1.

Type: Primary
Antigen: HIP12
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 451-550/1068
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge