Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDK5RAP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9076R-HRP
BOSSBS-9076R-HRPEA 1350 EUR InStock
BOSSBS-9076R-HRP
Anti-CDK5RAP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Cyclin dependent kinase 5 (Cdk5) is a key regulator of cell cycle progression in neuronal differentiation that physically associates with and is activated by the neuron-specific protein p35. CDK5RAP1 (CDK5 regulatory subunit-associated protein 1) specifically inhibits Cdk5 activation by p35 through formation of a dimer that inhibits kinase activity. CDK5RAP2, also known as Centrosome-associated protein 215, is a 1893 amino acid centrosomal protein that regulates activity of CDK5 through complex formation with CDK5RAP1. Expressed in placenta, liver, pancreas, heart, skeletal muscle, lung, brain and kidney, CDK5RAP2 associates with centrosomes throughout the cell cycle. Mutations in the gene encoding CDK5RAP2 results in primary microencephaly autosomal recessive type 3, which is characterized by markedly reduced head size, brain weight and significant neurological deficits. There are four isoforms of CDK5RAP2 that are produced as a result of alternative splicing events.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: CDK5RAP2
Clonality: Polyclonal
Clone: 0
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge