Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CINP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9077R-A680
BOSSBS-9077R-A680EA 893 EUR InStock
BOSSBS-9077R-A680
Anti-CINP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Cell cycle progression is controlled in part by a family of cyclin proteins and cyclin dependent kinases (Cdks). Cdk proteins work in concert with the cyclins to phosphorylate key substrates involved in each phase of cell cycle progression. Specifically, Cdk2 interacts with Cyclins A, B1, B3, D, or E to control cell cycle progression. The Cyclin-dependent kinase 2-interacting protein (CINP) interacts with components of the replication complex and Cdk2 and Cdc7, thereby providing a functional and physical link between Cdk2 and Cdc7 during firing of the origins of replication. However, CINP is phopshorylated by Cdc7, but not by Cdk2. CINP also interacts with ATR-interacting protein and regulates ATR-dependent signaling, resistance to replication stress and G2 checkpoint integrity.

Type: Primary
Antigen: CINP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/212
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge