Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ANKS1B Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9097R
BOSSBS-9097REA 725 EUR InStock
BOSSBS-9097R
Anti-ANKS1B Rabbit Polyclonal Antibody
Antikörper
The β-Amyloid protein precursor (AbPP) is a widely expressed transmembrane protein that is processed into the b-Amyloid (Ab) peptide, which accumulates in insoluble plaques in the brain of Alzheimer’s disease patients and AbPP intracellular domain (AID). AID may function as a pro-apoptotic peptide, a regulator of calcium homeostasis and a molecule involved in transcriptional regulation. The AID associated protein 1 (AIDA-1) is highly expressed in the brain and is regulated by AbPP. It interacts with AbPP to play a role in brain development. AIDA-1 also interacts with coilin in Cajal bodies to regulate pre-mRNA splicing.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ANKS1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge