Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ARSH Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9101R-CY3
BOSSBS-9101R-CY3EA 900 EUR InStock
BOSSBS-9101R-CY3
Anti-ARSH Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Sulfatases hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling and degradation of macromolecules. Arylsulfatase H, also known as ARSH, is a 562 amino acid protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Localized to the plasma membrane, Arylsulfatase H uses calcium as a cofactor to hydrolyze sulfate esters. The gene encoding Arylsulfatase D maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARSH
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge