Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ARTC5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9105R-A647
BOSSBS-9105R-A647EA 893 EUR InStock
BOSSBS-9105R-A647
Anti-ARTC5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Mono-ADP-ribosylation is one of the posttranslational protein modifications regulating cellular metabolism (e.g. nitrogen fixation) in prokaryotes. Mono-ADP-ribosylation is a process in which the ADP-ribose moiety of nicotinamide adenine dinucleotide is transferred to an acceptor amino acid. Five mammalian ADP-ribosyltransferases (ART1-ART5) have been cloned, and each ART is expressed in different tissues. ART5 (ADP-ribosyltransferase 5), also known as Ecto-ADP-ribosyltransferase 5, is a 292 amino acid secretory protein that is expressed in testis, heart, skeletal muscle and lymphoma. Functionally, ART5 is implicatetd to play a role in cell signaling and metabolism cascades. Two isoforms of ART5 exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARTC5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge