Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-POP1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9107R-CY3
BOSSBS-9107R-CY3EA 900 EUR InStock
BOSSBS-9107R-CY3
Anti-POP1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The death domain (DD) superfamily of proteins share one or more of the following domains: the DD, DED (death-effector domain), CARD (caspase-recruitment domain) and PYD (Pyrin domain). Each of these domains is characterized by a canonical death domain fold, which consists of a bundle of five or six antiparallel α-helices. As their names suggest, these domains play prominent roles in programmed cell death. ASC2 (apoptosis-associated speck-like protein containing a CARD 2), also known as Pyrin-only protein 1 or PADD-only protein 1, is an 89 amino acid member of the DD superfamily that contains one Pyrin domain. Localized to the cytoplasm, ASC2 interacts with ASC to modulate NF-κB and pro-caspase-1 regulation. ASC2 is predominantly expressed in monocytes, macrophages and granulocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: POP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge