Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BAGE3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9115R-A680
BOSSBS-9115R-A680EA 1350 EUR InStock
BOSSBS-9115R-A680
Anti-BAGE3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Unknown. Candidate gene encoding tumor antigens.Members of the BAGE gene family encode antigens that are recognised by cytotoxic T lymphocytes and are also known as CT (cancer/testis) antigens. Generated by juxtacentromeric shuffling of the MLL3 gene, the ancestral BAGE gene was expanded by acrocentric exchanges and/or juxtacentromeric movements.Generally, BAGE proteins are silent in all normal tissues with the exception of testis. BAGE2 and BAGE 3 (B melanoma antigen 2 and 3, respectively), also known as Cancer/testis antigen 2.2 and 2.3 (respectively), are 109 amino acid secreted proteins that are expressed in 22% of melanomas, lung and bladder carcinomas, and are also expressed in normal testis tissue. Like the genes encoding MAGE proteins, BAGE genes are most likely silenced by DNA methylation and/or chromatin compaction in normal tissues other than testis.

Type: Primary
Antigen: BAGE3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-109/109
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge