Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-INHBA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9116R-HRP
BOSSBS-9116R-HRPEA 900 EUR InStock
BOSSBS-9116R-HRP
Anti-INHBA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Members of the BAGE gene family encode antigens that are recognized by cytotoxic T lymphocytes and are also known as CT (cancer/testis) antigens. Generated by juxtacentromeric shuffling of the MLL3 gene, the ancestral BAGE gene was expanded by acrocentric exchanges and/or juxtacentromeric movements. BAGE, also known as B melanoma antigen 1 or cancer/testis antigen 2.1, is a 43 amino acid member of the BAGE family. Generally, BAGE proteins are not expressed in normal tissues, except in testis, but are expressed highly in melanomas, bladder carcinomas, head and neck squamous cell carcinomas, and lung and breast cancer carcinomas. BAGE proteins are not expressed in renal, colorectal and prostatic carcinomas, leukemias and lymphomas.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: INHBA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge