Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDCA7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9128R-A647
BOSSBS-9128R-A647EA 1350 EUR InStock
BOSSBS-9128R-A647
Anti-CDCA7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
CDCA7 is a nuclear protein without a known function, although its high homology with the transcription factor JPO2 or RAM2 suggests that it is also a transcription factor. CDCA7 is one of many target genes regulated by the c-Myc transcription factor. Overexpression of CDCA7 occurs in a significant proportion of cancers and it may play an important role in tumorigenesis. Normally it is periodically expressed in the cell cycle, peaking at the G1 to S phase transition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDCA7
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge