Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZNF230 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9143R-CY5
BOSSBS-9143R-CY5EA 900 EUR InStock
BOSSBS-9143R-CY5
Anti-ZNF230 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF141 (ring finger protein 141), also known as ZFP26 or ZNF230, is a 230 amino acid protein that contains one RING-type zinc finger. Expressed as two isoforms (isoform 1 and isoform 2) due to alternative splicing events, RNF141 is thought to function as a transcription factor during spermatogenesis. While isoform 2 is expressed in brain, heart, pancreas, kidney and skeletal muscle, isoform 1 is expressed primarily in testis, suggesting that isoform 1 functions during spermatogenesis. In addition, RNF141 is not expressed in azoospermic (infertile) men, further implicating an important role for RNF141 in testis development and male fertility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF230
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge