Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UBOX5 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9145R-CY3
BOSSBS-9145R-CY3EA 900 EUR InStock
BOSSBS-9145R-CY3
Anti-UBOX5 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The U-box domain is a modified RING finger motif that has been implicated in the ubiquitin/proteasome system. The ubiquitin-conjugating enzyme 7-interacting protein 5 (UIP5), also designated U-box domain-containing protein 5 or RING finger protein 37, contains 1 RING-type zinc finger and 1 U-box domain. UIP5 has been shown to interact with UBCH7, an enzyme that mediates selective degradation of abnormal proteins. The gene encoding UIP5 maps to chromosome 20, which houses over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome. Additionally, chromosome 20 contains a region with numerous genes which are thought important for seminal production and may be potential targets for male contraception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBOX5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge