Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRIM36 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9155R-CY7
BOSSBS-9155R-CY7EA 900 EUR InStock
BOSSBS-9155R-CY7
Anti-TRIM36 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
TRIM36 (tripartite motif-containing 36), also known as RNF98 (RING finger protein 98), HAPRIN (haploid germ cell-specific RBCC protein) or RBCC728, is a 728 amino acid protein that belongs to the TRIM/RBCC (Ring finger, B box, coiled-coil) family. Predominantly expressed in prostate, testis and brain with weak expression in heart, kidney and lung, TRIM36 contains two B box-type zinc fingers, a SPRY domain, a coiled-coil domain, a fibronectin type-III domain and a RING-type zinc finger; a motif that has zinc-chelating activity and is involved in mediating protein-protein and protein-DNA interactions. Localizing to the cytoplasm and the acrosomal region of germ cells and mature sperm, TRIM36 is believed to play a role in the acrosome reaction and fertilization. In addition, TRIM36 is overexpressed in prostate cancer, suggesting a possible role for TRIM36 in prostate tumorigenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRIM36
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge