Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF15 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9160R-A647
BOSSBS-9160R-A647EA 893 EUR InStock
BOSSBS-9160R-A647
Anti-RNF15 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
RORET, also known as RING finger protein 15 (RNF15) or zinc finger protein RoRet, is a 465 amino acid member of the TRIM family, also known as the RING-B-box coiled-coil (RBCC) family. Members of the RBCC family have an N-terminal RING finger, followed by one or two zinc-binding domains (B-box domains), a leucine coiled-coil region and a variable C-terminal domain. Found in all eukaryotes, members of the RBCC family typically function within a larger protein complex and possess ubiquitin-protein isopeptide ligase activity.The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF15
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge