Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRIM7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9164R-FITC
BOSSBS-9164R-FITCEA 900 EUR InStock
BOSSBS-9164R-FITC
Anti-TRIM7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The tripartite motif (TRIM) family of proteins are characterized by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. TRIM7 (tripartite motif-containing 7), also known as RNF90 or GNIP, is a 511 amino acid protein that belongs to the TRIM family and contains one RING-type zinc finger, one B box-type zinc finger and one SPRY domain. Expressed in placenta and skeletal muscle and present at lower levels in brain, heart and pancreas, TRIM7 localizes to both the cytoplasm and the nucleus where it exists as dimers and is thought to participate in the initiation of glycogen synthesis. Multiple isoforms of TRIM7 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRIM7
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge