Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9173R-A350
BOSSBS-9173R-A350EA 893 EUR InStock
BOSSBS-9173R-A350
Anti-RNF8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The RING finger motif is a specialized DNA-binding zinc finger domain found in many transcriptional regulatory proteins. The ring finger protein (RNF) family includes any protein containing the signature RING finger motif. RNF8 is a ubiquitously expressed nuclear RING finger protein that acts as an E3 ubiquitin-protein ligase. It is required for the ubiquitination of some nuclear proteins and promotes their subsequent degradation. The heterodimeric ubiquitin-conjugating enzyme UBC13 interacts with RNF8, and they co-localize in the nucleus. RNF8 may regulate mediation of UBC13 polyubiquitylation by elongating the ubiquitin chains. RNF8 also binds to Retinoid X receptor alpha (RXR?, a member of the steroid hormone receptor superfamily. It increases RXR?mediated transactivation of the RXR?responsive element (RXRE) promoter in a dose-dependent manner, suggesting that RNF8 is a regulator of RXR?mediated transcriptional activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge