Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRIM3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9174R-CY5.5
BOSSBS-9174R-CY5.5EA 900 EUR InStock
BOSSBS-9174R-CY5.5
Anti-TRIM3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Tripartite motif-containing protein 3 (TRIM3), also known as RING finger protein 22 (RNF22), RING finger protein 97 (RNF97) or brain-expressed RING finger protein (BERP), is a 744 amino acid member of the TRIM family, also known as the RING-B-box coiled-coil (RBCC) family. Members of the RBCC family have an N-terminal RING finger, followed by one or two zinc-binding domains (B-box domains), a leucine coiled-coil region and a variable C-terminal domain. Localized to cytoplasmic filaments, TRIM3 has been shown to interact with å-actinin-4 and myosin V, two proteins associated with the actin cytoskeleton. Specifically, å-actinin-4 interacts with the RBCC domain of TRIM3, and the C-terminal tail of Myosin V interacts with with the unique C-terminal ∫-propeller domain of TRIM3. These associations suggest that TRIM3 may play a role in cell motility and cargo transport. Three named isoforms of TRIM3 exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRIM3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge