Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Repetin Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9181R-A680
BOSSBS-9181R-A680EA 907 EUR InStock
BOSSBS-9181R-A680
Anti-Repetin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Repetin is a 784 amino acid secreted protein that belongs to the S100-fused protein family. Repetin is an extracellular epidermal matrix protein that is expressed in the epidermis and at high levels in eccrine sweat glands, the inner sheaths of hair roots and the filiform papilli of the tongue. A multifunctional epidermal matrix protein, repetin is involved in the cornified cell envelope formation. Containing two EF-hand domains, repetin?s recombinant N-terminal domain binds calcium reversibly. Repetin and trichohyalin may compensate for absent loricrin. The repetin gene contains three exons, two introns and spans at least 5.6 kb. The repetin gene is conserved in cow, mouse and rat, and maps to human chromosome 1q21.3.

Type: Primary
Antigen: Repetin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-140/784
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge