Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VGLL2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9183R-CY5.5
BOSSBS-9183R-CY5.5EA 900 EUR InStock
BOSSBS-9183R-CY5.5
Anti-VGLL2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Vgl-2, also known as VITO-1, is a 317 amino acid protein that contains a domain through which it interacts with TEF-1, a protein that plays a role in controlling the expression of numerous genes. Specific to skeletal muscle, Vgl-2 is expressed highly in adult fast muscle and is expressed at lower levels in adult slow muscle and fetal skeletal muscle. During muscle differentiation, Vgl-2 mRNA levels increase and Vgl-2 translocates from the cytoplasm to the nucleus. Overexpression of Vgl-2 in MYOD-transfected 10T1/2 mouse embryonic fibroblasts increases expression of myosin heavy chain (MHC), which is a marker of terminal muscle differentiation. This evidence suggests that Vgl-2 is essential for muscle gene expression. There are two isoforms of Vgl-2 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VGLL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge