Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VGLL3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9184R-CY5
BOSSBS-9184R-CY5EA 900 EUR InStock
BOSSBS-9184R-CY5
Anti-VGLL3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The transcriptional enhancer factor-1 (TEF-1) family of transcription factors regulate tissue-specific gene expression in muscle and placenta. The mechanism whereby TEF-1 confers tissue specificity depends largely on the interaction of TEF-1 with tissue-specific cofactors. Transcription cofactor Vgl-3 (vestigial-like protein 3), also known as colon carcinoma related protein, is a 326 amino acid nuclear protein that may act as a specific coactivator for the mammalian transcription elongation factors. Both Vgl-1 and Vgl-3 are enriched in placenta, whereas Vgl-2 is expressed in differentiating somites and branchial arches during embryogenesis and is skeletal-muscle specific in adult tissues. There are two isoforms of Vgl-3 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VGLL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge