Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZEB2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9186R-A555
BOSSBS-9186R-A555EA 893 EUR InStock
BOSSBS-9186R-A555
Anti-ZEB2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
SMAD regulates gene expression by interacting with different classes of transcription factors including DNA-binding multi-zinc finger proteins. SIP1, for SMAD interacting protein 1, is a member of the delta-EF1/Zfh1 family of 2-handed zinc finger/homeodomain proteins. SIP1 contains a SMAD-binding domain, a homeodomain and two clusters of zinc fingers on the N- and C-termini. SIP1, also known as SMADIP1, ZFHX1B and ZEB2 (zinc finger E-box-binding protein 2), can be induced by TGF∫ treatment. SIP1 plays a crucial role in normal embryonic development of neural structures and the neural crest. The human SIP1 gene maps to chromosome 2q22. Mutations in the SIP1 gene cause a form of Hirschsprung disease (HSCR). Patients with SIP1 mutations show mental retardation, delayed motor development, epilepsy, microcephaly, distinct facial features and/or congenital heart disease—all symptoms of HSCR.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZEB2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge