Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLFN14 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9190R-CY3
BOSSBS-9190R-CY3EA 900 EUR InStock
BOSSBS-9190R-CY3
Anti-SLFN14 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Schlafen family members are preferentially expressed in lymphoid tissues and are differentially regulated during thymocyte maturation. Schlafen proteins function as suppressors of cell growth and are thought to play a role in the maintenance of T cell quiescence. The prototype member of the Schlafen family, Slfn1, is transcriptionally unregulated during thymocyte positive selection, and the induction of Slfn1 induces a G0/G1 arrest, suggesting that Slfn1 participates in the regulation of cell cycle and potentially acts as a determining factor for apoptosis. These proteins all contain a largely conserved core domain within the center of the sequence, and yet they are substantially diversified at the N terminus. Slfn14 (Schlafen family member 14) is a 912 amino acid protein belonging to the Schlafen family. Slfn14 exists as two alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 17q12.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLFN14
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge