Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IGSF1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9194R-CY5
BOSSBS-9194R-CY5EA 900 EUR InStock
BOSSBS-9194R-CY5
Anti-IGSF1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
IGSF1 is a highly glycolylated immunoglobulin domain-containing protein. IGSF1 has been shown to act as a coreceptor in inhibin signaling, however, it does not appear to be a high-affinity inhibin receptor by itself. May reduce or inhibit activin A signaling and is believed to be necessary in the mediation of specific effects of inhibin B on activin-stimulated transcription. IGSF1 has been found to interact with several members of the ACVR family and possibly some members of the BMPR group. There are 3 known isoforms of IGSF1, with 1 and 2 likely being multi-pass membrane proteins. Isoform 3 is believed to be expressed as a secreted form. Expression is high in pancreas, testis and fetal liver, while heart, prostate and small intestine show only moderate expression. IGSF1 may be found at very low levels in brain, muscle, thymus, ovary, colon, fetal lung and fetal kidney. Isoform 3 has been detected in pituitary gland.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGSF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge