Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IGSF3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9196R-HRP
BOSSBS-9196R-HRPEA 900 EUR InStock
BOSSBS-9196R-HRP
Anti-IGSF3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
IGSF3, also known as V8 or EWI-3, is a 1,214 amino acid protein. Widely expressed with predominant expression in kidney, placenta and lung, IGSF3 localizes to the membrane and contains an N-terminal signal peptide, eight immunoglobulin (Ig) domains and a transmembrane segment. IGSF3 exhibits strong sequence and structural similarity to CD101 (32% identity), a leukocyte surface protein with seven Ig domains that is believed to play a role in T-cell activation. Despite the structural similarities between IGSF3 and CD101, IGSF3 is not expressed in peripheral blood lymphocytes and does not appear to participate in an immune function. Based on its subcellular localization and the presence of the eight Ig domains, IGSF3 is hypothesized to function as a surface receptor or as a cell adhesion molecule.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: IGSF3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge