Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SERPINA10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9209R-FITC
BOSSBS-9209R-FITCEA 900 EUR InStock
BOSSBS-9209R-FITC
Anti-SERPINA10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
ZPI, also known as SERPINA10 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10) or PZI, is a 444 amino acid secreted protein that functions as a Protein Z-dependent protease inhibitor. Expressed by the liver, ZPI is secreted into the plasma where, in the presence of calcium, Protein Z and phospholipids, it inhibits the activated pro-coagulation factors X and XI (Factor X and Factor XI). This inhibition helps properly regulate intravenous blood clotting. ZPI, a member of the serpin protein family, contains five potential N-linked glycosylation sites and a tyrosine at position 387 which, when disrupted, renders ZPI inactive. Defects in the gene encoding ZPI may increase susceptibility to venous thrombosis, the formation of blood clots within a vein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERPINA10
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge