Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF180 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9233R-A750
BOSSBS-9233R-A750EA 893 EUR InStock
BOSSBS-9233R-A750
Anti-RNF180 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF180 (ring finger protein 180), also known as Rines, is a 592 single-pass membrane protein that contains a single RING-type zinc finger. Expressed as three alternatively spliced isoforms, RNF180 is well conserved among vertebrates. RNF180 is expressed in brain, kidney, testis and uterus and localises to the endoplasmic reticulum. RNF180 is an E3 ubiquitin ligase involved in the ubiquitin-proteasome pathway. RNF180 is encoded by a gene located on human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm, or of chromosome 5 altogether, is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.

Type: Primary
Antigen: RNF180
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/592
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge