Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF23 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9238R-A350
BOSSBS-9238R-A350EA 893 EUR InStock
BOSSBS-9238R-A350
Anti-RNF23 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF23 (RING finger protein 23), also known as tripartite motif-containing protein 39 (TRIM39) or testis-abundant finger protein, is a 518 amino acid protein belonging to the TRIM/RBCC family that is known to interact with MOAP1. Ubiquitously expressed and existing as two alternatively spliced isoforms, RNF23 is found at highest levels in spleen, testis, brain, kidney, liver, heart and skeletal muscle. RNF23 typically localizes to cytosol but shifts to mitochondria upon co-localization with MOAP1, a short-lived, pro-apoptotic protein which RNF23 prevents from becoming poly-ubiquitinated and degraded, thereby facilitating apoptosis. RNF23 contains one B box-type zinc finger, a B30.2/SPRY domain and a single RING-type zinc finger.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF23
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge