Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF21 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9244R-CY5.5
BOSSBS-9244R-CY5.5EA 900 EUR InStock
BOSSBS-9244R-CY5.5
Anti-RNF21 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Tripartite motif-containing protein 34 (TRIM34), also known as RING finger protein 21 (RNF21) or interferon-responsive finger protein 1 (IFP1), is a 488 amino acid member of the TRIM family, also known as the RING-B-box coiled-coil (RBCC) family. Members of the RBCC family have an N-terminal RING finger, followed by one or two zinc-binding domains (B-box domains), a leucine coiled-coil region and a variable C-terminal domain. Three isoforms of TRIM34 exist as a result of alternative splicing events. Isoform 1, the most abundant isoform, is highly expressed in placenta, spleen, colon and peripheral blood leukocytes. Studies have shown that Interferon (IFN) stimulation leads to an upregulation of TRIM34. These findings suggest that TRIM34 maybe a downstream effector that mediates IFN activities.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF21
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge