Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF113A Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9248R-CY3
BOSSBS-9248R-CY3EA 900 EUR InStock
BOSSBS-9248R-CY3
Anti-RNF113A Rabbit Polyclonal Antibody (Cy3®)
Antikörper
RNF113A is a novel gene whose function cannot directly be inferred from it's sequence analysis. Ring finger proteins have a role in signalling the destruction of a range of heterologous protein substrates. They are small zinc binding domains present within arrays of larger, functionally distinct proteins, often close to the amino or carboxyl termini. RNF113A is a ubiquitously expressed protein that contains a RING type zinc finger and a C3H1 type zinc finger.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF113A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge