Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF130 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9252R-A350
BOSSBS-9252R-A350EA 893 EUR InStock
BOSSBS-9252R-A350
Anti-RNF130 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF130 (ring finger protein 130), also known as GP, G1RZFP (G1-related zinc finger protein) or GOLIATH, is a 419 amino acid single-pass type I membrane protein that shares similarity with a Drosophila zinc-finger protein found in mesoderm known as g1. RNF130 contains one PA (protease associated) domain and a single RING-type zinc finger. Implicated in the regulation of growth factor withdrawal-induced apoptosis of myeloid precursor cells, RNF130 is encoded by a gene located on human chromosome 5q35.3 and mouse chromosome 11 B1.3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF130
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge