Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF165 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9258R-CY3
BOSSBS-9258R-CY3EA 900 EUR InStock
BOSSBS-9258R-CY3
Anti-RNF165 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in protein-protein interactions and protein-DNA interactions. Specifically, they are thought to be involved in the ubiquitination pathway of protein degradation. RNF165 (RING finger protein 165) is a 346 amino acid protein that contains one RING-type zinc finger. Via its RING-type zinc finger, RNF165 may play a role in transcriptional regulation and protein degradation events. The gene encoding RNF165 maps to human chromosome 18, which houses over 300 genes, including Trisomy 18, or Edwards syndrome, the second most common trisomy after Downs syndrome. Translocation between chromosome 18 and 14 is the most common translocation in cancers, and occurs in follicular lymphomas. Niemann-Pick disease, hereditary hemorrhagic telangiectasia and erythropoietic protoporphyria are also associated with chromosome 18.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF165
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge