Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF131 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9290R-CY3
BOSSBS-9290R-CY3EA 900 EUR InStock
BOSSBS-9290R-CY3
Anti-RNF131 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
PJA2, also known as E3 ubiquitin-protein ligase praja-2, RNF131 (ring finger protein 131) or Neurodap1, is a 708 amino acid protein that contains one ring-type zinc finger and exists as two alternatively spliced isoforms. Significantly conserved in chimpanzee, dog, cow, mouse, rat, chicken and zebrafish, PJA2 shares 52% identity with PJA1, which is involved in protein ubiquitination in brain and may play a role in X-linked mental retardation. Encoded by a gene that maps to human chromosome 5q21.3, PJA2 localizes to both endoplasmic reticulum and Golgi apparatus membranes. Participating in E2-dependent, E3 ubiquitin-protein ligase activity, PJA2 binds to a variety of E2s and interacts with ubiquitin-conjugating enzymes, such as UBE2D2, in vitro.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF131
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge