Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NNP1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9300R-CY3
BOSSBS-9300R-CY3EA 900 EUR InStock
BOSSBS-9300R-CY3
Anti-NNP1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Novel nuclear protein 1 (NNP-1), also known as RRP1-like protein or Nucleolar protein Nop52, is a 461 amino acid protein belonging to the RRP1 family. Localized to the nucleolus, NNP-1 has simian virus 40-type and bipartite nuclear localization signals and four coiled-coil domains within its C-terminal region. NNP-1 has been found to play an important role in the generation of 28S rRNA in the late processing steps of ribosome biogenesis. At the end of mitosis, nucleolar proteins assemble in a sequential order during the rebuilding of the nucleolus. NNP-1 assembles after Fibrillarin and C23, and simultaneously with B23 and POP1 in the prenucleolar body pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NNP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge