Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SPO11 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9302R-CY3
BOSSBS-9302R-CY3EA 900 EUR InStock
BOSSBS-9302R-CY3
Anti-SPO11 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Spo11 is a type II topoisomerase that is thought to generate the chromosome breaks that initiate meiotic recombination. The Spo11 protein initiates meiotic recombination by generating DNA double-strand breaks (DSBs) and is required for meiotic synapsis in S. cerevisiae. The DSBs are located mostly in promoter regions, where the chromatin is in an open configuration, and cluster in domains along the chromosome. Expression of the Spo11 is detected mainly in the testis, in agreement with its predicted function in the initiation of meiotic recombination. Disruption of Spo11 leads to severe gonadal abnormalities from defective meiosis and results in infertility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPO11
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge