Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KATNA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9308R-A647
BOSSBS-9308R-A647EA 893 EUR InStock
BOSSBS-9308R-A647
Anti-KATNA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Microtubules are polymers of alpha and beta subunits that form the mitotic spindle and assist in the organization of membranous organelles during interphase. Katanin p60 A1, also known as KATNA1, is a 491 amino acid protein that belongs to the AAA ATPase family and is involved in microtubule regulation. Localized to the cytoplasm and to the centrosome, Katanin p60 A1 functions to sever and disassemble microtubules in an ATP-dependent manner, thus promoting the rapid reorganization of cellular microtubule arrays and playing an important role in microtubule release from the centrosome after nucleation. Katanin p60 A1, which exists as two alternatively spliced isoforms, can homooligomerize into hexameric rings whose activity is stimulated by the presence of microtubules.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KATNA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge